Assay Details
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012909.1 | NP_001012927.1 | 11 - 12 | 1459 | 65 | |
XM_015288632.2 | XP_015144118.1 | 11 - 12 | 1494 | 65 | ||
GenBank mRNA | AJ851690.1 | - | 11 - 12 | 1459 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
