Assay Details
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012909.1 | NP_001012927.1 | 3 - 4 | 420 | 95 | |
XM_015288632.2 | XP_015144118.1 | 3 - 4 | 455 | 95 | ||
GenBank mRNA | AJ851690.1 | - | 3 - 4 | 420 | 95 | |
CR353263.1 | - | 1 - 2 | 73 | 95 | ||
CR387792.1 | - | 2 - 3 | 108 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
