Assay Details
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012909.1 | NP_001012927.1 | 4 - 5 | 626 | 115 | |
XM_015288632.2 | XP_015144118.1 | 4 - 5 | 115 | |||
GenBank mRNA | AJ851690.1 | - | 4 - 5 | 626 | 115 | |
CR353263.1 | - | 2 - 3 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
