Assay Details
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012941.2 | NP_001012959.1 | 1 - 2 | 256 | 71 | |
GenBank mRNA | AB124571.1 | - | 1 - 2 | 116 | 71 | |
AB196971.1 | - | 1 - 2 | 256 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
