Assay Details
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012941.2 | NP_001012959.1 | 3 - 4 | 968 | 73 | |
GenBank mRNA | AB124571.1 | - | 3 - 4 | 828 | 73 | |
AB196971.1 | - | 3 - 4 | 968 | 73 | ||
CR407279.1 | - | 2 - 3 | 646 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
