Assay Details
Gene Symbol: RHOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029849.1 | NP_001025020.1 | 1 | 432 | 60 | |
GenBank mRNA | AF098514.1 | - | 1 | 432 | 60 | |
U79759.1 | - | 349 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
