Assay Details
Gene Symbol: OSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029981.2 | NP_001025152.1 | 2 - 3 | 158 | 80 | |
XM_015296367.2 | XP_015151853.1 | 2 - 3 | 208 | 80 | ||
XM_015296368.2 | XP_015151854.1 | 2 - 3 | 152 | 80 | ||
XM_015296369.2 | XP_015151855.1 | 2 - 3 | 180 | 80 | ||
XM_015296370.2 | XP_015151856.1 | 2 - 3 | 174 | 80 | ||
XM_015296371.2 | XP_015151857.1 | 2 - 3 | 1071 | 80 | ||
XM_015296372.2 | XP_015151858.1 | 2 - 3 | 1080 | 80 | ||
XM_025142197.1 | XP_024997965.1 | 2 - 3 | 151 | 80 | ||
GenBank mRNA | BX930492.2 | - | 2 - 3 | 158 | 80 | |
BX931834.1 | - | 2 - 3 | 161 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
