Assay Details
Gene Symbol: RGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031216.1 | NP_001026387.1 | 5 - 6 | 691 | 78 | |
GenBank mRNA | AY339627.1 | - | 5 - 6 | 691 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
