Assay Details
Gene Symbol: FRZB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204772.2 | NP_990103.1 | 1 - 2 | 497 | 73 | |
GenBank mRNA | AF153476.1 | - | 1 - 2 | 322 | 73 | |
DQ017062.1 | - | 1 - 2 | 497 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
