Assay Details
Gene Symbol: FRZB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204772.2 | NP_990103.1 | 2 - 3 | 545 | 87 | |
GenBank mRNA | AF153476.1 | - | 2 - 3 | 370 | 87 | |
AF207665.1 | - | 2 - 3 | 390 | 87 | ||
DQ017062.1 | - | 2 - 3 | 545 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
