Assay Details
Gene Symbol: FRZB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204772.2 | NP_990103.1 | 3 - 4 | 606 | 93 | |
GenBank mRNA | AF218057.1 | - | 3 - 4 | 603 | 93 | |
DQ017062.1 | - | 3 - 4 | 606 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
