Assay Details
Gene Symbol: FRZB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204772.2 | NP_990103.1 | 4 - 5 | 811 | 102 | |
GenBank mRNA | AF218057.1 | - | 4 - 5 | 808 | 102 | |
DQ017062.1 | - | 4 - 5 | 811 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
