Assay Details
Gene Symbol: GART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001469.1 | NP_001001469.1 | 11 - 12 | 1344 | 87 | |
XM_015299217.2 | XP_015154703.1 | 13 - 14 | 1553 | 87 | ||
XM_015299227.1 | XP_015154713.1 | 14 - 15 | 1666 | 87 | ||
XM_015299230.2 | XP_015154716.1 | 13 - 14 | 1606 | 87 | ||
XM_015299235.2 | XP_015154721.1 | 12 - 13 | 1509 | 87 | ||
XM_025153908.1 | XP_025009676.1 | 13 - 14 | 1636 | 87 | ||
GenBank mRNA | FJ977573.1 | - | 1 - 2 | 191 | 87 | |
X54200.1 | - | 1344 | 87 | |||
X56339.1 | - | 1344 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
