Assay Details
Gene Symbol: GART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001469.1 | NP_001001469.1 | 18 - 19 | 2486 | 96 | |
XM_015299217.2 | XP_015154703.1 | 20 - 21 | 2695 | 96 | ||
XM_015299227.1 | XP_015154713.1 | 21 - 22 | 2808 | 96 | ||
XM_015299230.2 | XP_015154716.1 | 20 - 21 | 2748 | 96 | ||
XM_015299235.2 | XP_015154721.1 | 19 - 20 | 2651 | 96 | ||
XM_025153908.1 | XP_025009676.1 | 20 - 21 | 2778 | 96 | ||
GenBank mRNA | CR387893.1 | - | 2 - 3 | 256 | 96 | |
FJ977574.1 | - | 2 - 3 | 226 | 96 | ||
X54200.1 | - | 2486 | 96 | |||
X56339.1 | - | 2486 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
