Assay Details
Gene Symbol: GART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001469.1 | NP_001001469.1 | 20 - 21 | 2757 | 71 | |
XM_015299217.2 | XP_015154703.1 | 22 - 23 | 2966 | 71 | ||
XM_015299227.1 | XP_015154713.1 | 23 - 24 | 3079 | 71 | ||
XM_015299230.2 | XP_015154716.1 | 22 - 23 | 3019 | 71 | ||
XM_015299235.2 | XP_015154721.1 | 21 - 22 | 2922 | 71 | ||
XM_025153908.1 | XP_025009676.1 | 22 - 23 | 3049 | 71 | ||
GenBank mRNA | FJ977574.1 | - | 4 - 5 | 497 | 71 | |
X54200.1 | - | 2757 | 71 | |||
X56339.1 | - | 2757 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
