Assay Details
Gene Symbol: LOC414835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001754.1 | NP_001001754.1 | 13 - 14 | 3109 | 99 | |
XM_015282913.2 | XP_015138399.1 | 13 - 14 | 7862 | 99 | ||
XM_015282914.2 | XP_015138400.1 | 13 - 14 | 7862 | 99 | ||
XM_025147081.1 | XP_025002849.1 | 10 - 11 | 1753 | 99 | ||
GenBank mRNA | AY312363.1 | - | 13 - 14 | 3109 | 99 | |
AY355452.1 | - | 1 - 2 | 97 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
