Assay Details
Gene Symbol: LOC414835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001754.1 | NP_001001754.1 | 26 - 27 | 6845 | 63 | |
XM_015282913.2 | XP_015138399.1 | 26 - 27 | 11598 | 63 | ||
XM_015282914.2 | XP_015138400.1 | 26 - 27 | 11598 | 63 | ||
XM_025147081.1 | XP_025002849.1 | 23 - 24 | 5489 | 63 | ||
GenBank mRNA | AY312363.1 | - | 26 - 27 | 6845 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
