Assay Details
Gene Symbol: MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001782.1 | NP_001001782.1 | 10 - 11 | 1677 | 125 | |
XM_015290974.2 | XP_015146460.1 | 9 - 10 | 2530 | 125 | ||
GenBank mRNA | AY613435.1 | - | 10 - 11 | 1677 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
