Assay Details
Gene Symbol: MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001782.1 | NP_001001782.1 | 2 - 3 | 238 | 69 | |
XM_025152796.1 | XP_025008564.1 | 2 - 3 | 1016 | 69 | ||
GenBank mRNA | AY613435.1 | - | 2 - 3 | 238 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
