Assay Details
Gene Symbol: MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001782.1 | NP_001001782.1 | 6 - 7 | 941 | 95 | |
XM_015290974.2 | XP_015146460.1 | 6 - 7 | 1728 | 95 | ||
XM_015290975.2 | XP_015146461.1 | 6 - 7 | 1728 | 95 | ||
XM_025152794.1 | XP_025008562.1 | 6 - 7 | 1728 | 95 | ||
XM_025152795.1 | XP_025008563.1 | 6 - 7 | 1728 | 95 | ||
XM_025152796.1 | XP_025008564.1 | 6 - 7 | 1719 | 95 | ||
XM_025152797.1 | XP_025008565.1 | 6 - 7 | 1728 | 95 | ||
XM_025152799.1 | XP_025008567.1 | 6 - 7 | 1728 | 95 | ||
XM_025152800.1 | XP_025008568.1 | 6 - 7 | 1728 | 95 | ||
XM_025152801.1 | XP_025008569.1 | 6 - 7 | 1728 | 95 | ||
XM_025152802.1 | XP_025008570.1 | 6 - 7 | 1728 | 95 | ||
GenBank mRNA | AY613435.1 | - | 6 - 7 | 941 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
