Assay Details
Gene Symbol: MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001782.1 | NP_001001782.1 | 8 - 9 | 1263 | 76 | |
XM_025152797.1 | XP_025008565.1 | 8 - 9 | 2050 | 76 | ||
GenBank mRNA | AY613435.1 | - | 8 - 9 | 1263 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
