Assay Details
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004379.1 | NP_001004379.1 | 3 - 4 | 418 | 113 | |
GenBank mRNA | CR385365.1 | - | 3 - 4 | 423 | 113 | |
X62640.1 | - | 418 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
