Assay Details
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005791.1 | NP_001005791.1 | 2 - 3 | 462 | 93 | |
GenBank mRNA | AJ583692.2 | - | 2 - 3 | 459 | 93 | |
AJ719832.1 | - | 2 - 3 | 462 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
