Assay Details
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005791.1 | NP_001005791.1 | 4 - 5 | 627 | 111 | |
GenBank mRNA | AJ583692.2 | - | 4 - 5 | 624 | 111 | |
AJ719832.1 | - | 4 - 5 | 627 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
