Assay Details
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006415.1 | NP_001006415.1 | 5 - 6 | 892 | 125 | |
XM_025148781.1 | XP_025004549.1 | 5 - 6 | 929 | 125 | ||
GenBank mRNA | AJ719453.1 | - | 5 - 6 | 892 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
