Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204376.2 | NP_989707.1 | 3 - 4 | 328 | 65 | |
GenBank mRNA | AJ720701.1 | - | 3 - 4 | 328 | 65 | |
AY336970.1 | - | 3 - 4 | 328 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
