Assay Details
Gene Symbol: CXCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204712.2 | NP_990043.1 | 2 - 3 | 281 | 64 | |
GenBank mRNA | AF285876.2 | - | 2 - 3 | 281 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
