Assay Details
Gene Symbol: MRPS30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204939.1 | NP_990270.1 | 1 - 2 | 743 | 77 | |
GenBank mRNA | AF029071.1 | - | 1 - 2 | 743 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
