Assay Details
Gene Symbol: GFRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204991.1 | NP_990322.1 | 2 - 3 | 313 | 114 | |
XM_015286040.2 | XP_015141526.1 | 2 - 3 | 340 | 114 | ||
GenBank mRNA | AF045162.1 | - | 2 - 3 | 313 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
