Assay Details
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205017.1 | NP_990348.1 | 3 - 4 | 241 | 66 | |
GenBank mRNA | U79755.1 | - | 3 - 4 | 241 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
