Assay Details
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205017.1 | NP_990348.1 | 4 - 5 | 302 | 107 | |
XM_015294138.2 | XP_015149624.1 | 5 - 6 | 502 | 107 | ||
GenBank mRNA | U79755.1 | - | 4 - 5 | 302 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
