Assay Details
Gene Symbol: NES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205033.2 | NP_990364.2 | 1037 | 93 | ||
GenBank mRNA | U59287.2 | - | 1037 | 93 | ||
X80877.1 | - | 3 - 4 | 1039 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
