Assay Details
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205044.2 | NP_990375.1 | 2 - 3 | 185 | 85 | |
GenBank mRNA | Y13903.1 | - | 185 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
