Assay Details
Gene Symbol: AKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205055.1 | NP_990386.1 | 2 - 3 | 511 | 70 | |
GenBank mRNA | AF039943.1 | - | 2 - 3 | 511 | 70 | |
BX930420.1 | - | 2 - 3 | 336 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
