Assay Details
Gene Symbol: GABRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205058.1 | NP_990389.1 | 9 - 10 | 1343 | 126 | |
GenBank mRNA | X94343.1 | - | 1343 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
