Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205292.1 | NP_990623.1 | 2 - 3 | 385 | 78 | |
GenBank mRNA | HQ439436.1 | - | 2 - 3 | 379 | 78 | |
X03578.1 | - | 2 - 3 | 420 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
