Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205292.1 | NP_990623.1 | 4 - 5 | 671 | 73 | |
GenBank mRNA | HQ439436.1 | - | 4 - 5 | 665 | 73 | |
X03578.1 | - | 4 - 5 | 706 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
