Assay Details
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205339.2 | NP_990670.2 | 1 - 2 | 737 | 72 | |
GenBank mRNA | Z11961.1 | - | 1 - 2 | 626 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
