Assay Details
Gene Symbol: UTS2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206989.2 | NP_996872.1 | 2 - 3 | 276 | 98 | |
GenBank mRNA | AY566611.1 | - | 2 - 3 | 276 | 98 | |
CR406384.1 | - | 2 - 3 | 270 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
