Assay Details
Gene Symbol: COX4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030577.1 | NP_001025748.1 | 2 - 3 | 118 | 89 | |
XM_015292558.1 | XP_015148044.1 | 2 - 3 | 209 | 89 | ||
GenBank mRNA | AJ720329.1 | - | 2 - 3 | 118 | 89 | |
BX930028.2 | - | 2 - 3 | 121 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
