Assay Details
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030619.1 | NP_001025790.1 | 2 - 3 | 188 | 66 | |
XM_015293784.2 | XP_015149270.1 | 2 - 3 | 474 | 66 | ||
XM_015293785.2 | XP_015149271.1 | 2 - 3 | 385 | 66 | ||
GenBank mRNA | AJ721016.1 | - | 2 - 3 | 188 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
