Assay Details
Gene Symbol: CBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030795.2 | NP_001025966.1 | 3 - 4 | 466 | 94 | |
XM_015299436.2 | XP_015154922.1 | 3 - 4 | 516 | 94 | ||
XM_025149473.1 | XP_025005241.1 | 3 - 4 | 456 | 94 | ||
GenBank mRNA | BX936209.2 | - | 2 - 3 | 357 | 94 | |
DQ079061.1 | - | 3 - 4 | 466 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
