Assay Details
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030804.1 | NP_001025975.1 | 4 - 5 | 422 | 152 | |
GenBank mRNA | AJ720842.1 | - | 4 - 5 | 422 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() |
