Assay Details
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030808.1 | NP_001025979.1 | 2 - 3 | 785 | 64 | |
GenBank mRNA | AJ851372.1 | - | 2 - 3 | 785 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
