Assay Details
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030825.2 | NP_001025996.2 | 9 - 10 | 1259 | 87 | |
GenBank mRNA | AJ720302.1 | - | 9 - 10 | 1257 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
