Assay Details
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030825.2 | NP_001025996.2 | 4 - 5 | 457 | 121 | |
GenBank mRNA | AJ720302.1 | - | 4 - 5 | 458 | 121 | |
CR523864.1 | - | 2 - 3 | 843 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
