Assay Details
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030930.1 | NP_001026101.1 | 7 - 8 | 700 | 104 | |
GenBank mRNA | AJ720035.1 | - | 7 - 8 | 700 | 104 | |
BX929898.2 | - | 4 - 5 | 439 | 104 | ||
EF426481.1 | - | 7 - 8 | 687 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
