Assay Details
Gene Symbol: E2F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030942.1 | NP_001026113.1 | 6 - 7 | 894 | 70 | |
XM_015282807.2 | XP_015138293.2 | 6 - 7 | 796 | 70 | ||
GenBank mRNA | AJ720953.1 | - | 6 - 7 | 894 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
