Assay Details
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030995.1 | NP_001026166.1 | 4 - 5 | 761 | 94 | |
XM_015281562.2 | XP_015137048.1 | 4 - 5 | 571 | 94 | ||
GenBank mRNA | AJ719290.1 | - | 4 - 5 | 790 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
