Assay Details
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031226.1 | NP_001026397.1 | 2 - 3 | 248 | 124 | |
GenBank mRNA | AJ719394.1 | - | 2 - 3 | 248 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
